R&D Systems Human LR3 IGF-I Recombinant Protein

Manufacturer: R&D Systems 8335G1200

 View more versions of this product

Catalog No. 83-35G-1200

  Check Availability
Add to Cart

    Specifications

    Quantity 200μg
    Termination Met-Phe-Pro-Ala-Met-Pro-Leu-Ser-Ser-Leu
    Without Additives Carrier Protein
    Gene ID (Entrez) 3479
    Preparation Method Reconstitute at 500μg/mL in sterile PBS.
    Purity 95%, by SDS-PAGE with silver staining
    Recombinant Yes
    Source E. coli-derived, Gly49-Ala118 (Glu51Arg), with an N-terminal tag
    Shelf Life 365 days
    Formulation Lyophilized from a 0.2μm filtered solution in phosphate buffer, pH 7.2
    Accession Number P05019
    Storage Requirements -20 to -70°C
    Name LR3 IGF-I
    Quality Control Testing SDS-PAGE: 7kDa, reducing conditions
    Regulatory Status RUO
    Conjugate Unlabeled
    Molecular Weight 9kDa

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit