Learn More
R&D Systems Human Lymphotoxin α1/β2 Recombinant Protein
Manufacturer:  R&D Systems 678LY010/CF/DELL
 View more versions of this product
Catalog No. 678LY010CF
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications Description & Specifications
Specifications
Lyophilized from a 0.2μm filtered solution in PBS.
18 kDa (LT alpha), 18.5 kDa (LTbeta)
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Unlabeled
4049
Lymphotoxin α1/β2
10μg
Noncovalently-linked heterotrimer
Yes