R&D Systems Human MBL Recombinant Protein

Manufacturer: R&D Systems 2307MB050

 View more versions of this product

Catalog No. 2307MB050

  Check Availability
Add to Cart

    Specifications

    Molecular Weight 24 kDa (monomer)
    Name MBL
    Recombinant Yes
    Structural Form Oligomer
    Conjugate Unlabeled
    Formulation Lyophilized from a 0.2μm filtered solution in PBS with Trehalose and BSA as a carrier protein.
    Gene ID (Entrez) 4153
    Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
    Quantity 50μg
    Source Human

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit