Learn More
R&D Systems Human MD-1 Recombinant Protein
Manufacturer:  R&D Systems 925MD050/CF
 View more versions of this product
Catalog No. 925MD050CF
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Lyophilized from a 0.2μm filtered solution in PBS.
32.5 kDa
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Yes
9450
MD-1
50μg
Unlabeled