R&D Systems Human MIS/AMH Recombinant Protein

Manufacturer: R&D Systems 1737MS010/CF

 View more versions of this product

Catalog No. 1737MS010CF

  Check Availability
Add to Cart

    Specifications

    Molecular Weight 11.7 kDa (monomer)
    Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
    Formulation Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
    Gene ID (Entrez) 268
    Name MIS/AMH
    Quantity 10μg
    Recombinant Yes
    Source Human
    Structural Form Disulfide-linked homodimer
    Conjugate Unlabeled

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit