Learn More
R&D Systems™ Human/Mouse/Rat/Canine/Equine BDNF Recombinant Protein Available on GSA/VA Contract for Federal Government customers only.
Manufacturer:  R&D Systems™ 248BD025
 View more versions of this product
Catalog No. 248BD025
Add to cart
This item is not returnable. View return policy
Description
Description
Measured in a cell proliferation assay using BaF mouse pro-B cells transfected with TrkB. The ED 50 for this effect is 0.2-2 ng/mL. The specific activity of Recombinant Human/Mouse/Rat/Canine/Equine BDNF is approximately 1.3 x 10 3 units/µg, which is calibrated against recombinant human BDNF WHO Standard (NIBSC code: 96/534). Measured by its binding ability in a functional ELISA. When Recombinant Human TrkB Fc Chimera (Catalog # 688-TK ) is coated at 1 µg/mL, Recombinant Human BDNF binds with an apparent K d <1 nM.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Lyophilized from a 0.2μm filtered solution in Sodium Citrate and NaCl with BSA as a carrier protein.
13.5 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Unlabeled
627
BDNF
25μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Yes