Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human/Mouse/Rat/Canine/Equine BDNF Recombinant Protein

Manufacturer:  R&D Systems™ 248BD025/CF

 View more versions of this product

Catalog No. 248BD025CF

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.

This item is not returnable. View return policy

Description

Description

Measured in a cell proliferation assay using BaF mouse pro-B cells transfected with TrkB. The ED 50 for this effect is 0.2-2 ng/mL. The specific activity of Recombinant Human/Mouse/Rat/Canine/Equine BDNF is approximately 1.3 x 10 3 units/µg, which is calibrated against recombinant human BDNF WHO Standard (NIBSC code: 96/534). Measured by its binding ability in a functional ELISA. When Recombinant Human TrkB Fc Chimera (Catalog # 688-TK ) is coated at 1 µg/mL, Recombinant Human BDNF binds with an apparent K d <1 nM.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in Sodium Citrate and NaCl.
13.5 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Unlabeled
627
BDNF
25μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Yes
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit