Learn More
R&D Systems Human MSP/MST1 β Chain Recombinant Protein
Manufacturer:  R&D Systems 635MS025/CF
 View more versions of this product
Catalog No. 635MS025CF
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
25 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Yes
4485
MSP/MST1 y Chain
25μg
Unlabeled