Learn More
R&D Systems Human MuRF1/TRIM63 Ubiquitin Ligase Kit, S5a Substrate
Designed for in vitro MuRF1-mediated ubiquitination of user-supplied substrates such as cardiac troponin 1, MYH7, or muscle creatine kinase
Manufacturer:  R&D Systems K102
Catalog No. K102
Add to cart
This item is not returnable. View return policy
Description
Description
MuRF1 (Muscle-specific RING-finger protein 1) is a RING-finger E3 ligase found in striated muscle (heart and skeletal) and iris tissues. The protein contains both RING and B-box zinc fingers, and a coiled-coil tripartite fold known as TRIM. MuRF1 ligase activity regulates the proteasomal degradation of cardiac troponin 1 and other sarcomeric-associated proteins.
Ubiquitinated proteins can be used in downstream applications, or analyzed by Western blot using antibodies specific for the target protein
Specifications
Specifications
Store the unopened product at -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Angiocidin, ASF, EC 6.3.2.-, IRF, IRFMURF2, Iris RING finger protein, Macropain, Mcb1, MuRF1, MuRF-1, muscle specific ring finger protein 2, Muscle-specific RING finger protein 1, PSMD4, pUB-R5, RING finger protein 28FLJ32380, RNF28, RNF28E3 ubiquitin-pro