Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Netrin-1 Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

$490.68

Specifications

Gene ID (Entrez) 9423
Molecular Weight (g/mol) 67.5 kDa
Name Netrin-1
Quantity 25μg
Source Mouse myeloma cell line,NS0-derived human Netrin-1 protein, (N-terminus) Human Netrin-1 (Val22-Ala604) Accession &Num; O95631 HPGGGSGGGSGGGS 6-His tag (C-terminus)
View More Specs

Products 2
Catalog Number Mfr. No. Formulation Price Quantity & Availability  
Catalog Number Mfr. No. Formulation Price Quantity & Availability  
6419N1025CF
View Documents
R&D Systems™
6419N1025/CF
Lyophilized from a 0.2μm filtered solution in PBS and EDTA.
Each for $490.68
Add to Cart
 
6419N1025
View Documents
R&D Systems™
6419N1025
Lyophilized from a 0.2μm filtered solution in PBS and EDTA with BSA as a carrier protein.
Each for $490.68
Add to Cart
 
Description

Description

The Recombinant Human Netrin-1 Protein is derived from NS0. The Recombinant Human Netrin-1 Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

9423
Netrin-1
Mouse myeloma cell line,NS0-derived human Netrin-1 protein, (N-terminus) Human Netrin-1 (Val22-Ala604) Accession &Num; O95631 HPGGGSGGGSGGGS 6-His tag (C-terminus)
Unconjugated
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
67.5 kDa
25μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Recombinant
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit