Learn More
R&D Systems™ Human NKp46/NCR1 Fc Chimera Recombinant Protein Available on GSA/VA Contract for Federal Government customers only.
Manufacturer:  R&D Systems™ 1850NK025
Catalog No. 1850NK025
Add to cart
This item is not returnable. View return policy
Description
Description
Measured by its ability to bind Recombinant Human Vimentin (Catalog # 2105-VI ) in a functional ELISA with an estimated K D < 2 nM. Garg, A. et al. (2006) J. Immunol. 177:6192.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Lyophilized from a 0.2μm filtered solution in PBS.
53 kDa (monomer)
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
NS0
Disulfide-linked homodimer
Yes
9437
NKp46/NCR1 Fc Chimera
25μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unlabeled