Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human Pappalysin-1/PAPP-A aa 82-1214 Recombinant Protein

Manufacturer:  R&D Systems™ 2487ZN020

Catalog No. 2487ZN020


Add to cart
This item is not returnable. View return policy

Description

Description

Source: Mouse myeloma cell line, NS0-derived, Glu82-Asp1214 and Arg84-Asp1214, both with a C-terminal 10-His tag
Purity: 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 128 kDa
Formulation: Supplied as a 0.2µm filtered solution in Tris, NaCl and CaCl2.
Entrez Gene ID: 5069
Regulatory Status: RUO

Specifications

Specifications

Supplied as a 0.2 μm filtered solution in Tris, NaCl and CaCl 2 .
Predicted MW: 128kDa Observed MW: 120kDa and 160kDa, reducing conditions
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Disulfide-linked homodimer
Yes
5069
Pappalysin-1/PAPP-A aa 82-1214
20μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening.
Unlabeled
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit