Learn More
R&D Systems™ Human PCPE-1 Recombinant Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Supplier:  R&D Systems™ 2627PE020
Catalog No. 2627PE020
Add to Cart
This item is not returnable. View return policy
Description
Description
The Recombinant Human PCPE-1 Protein is derived from NS0. The Recombinant Human PCPE-1 Protein has been validated for the following applications: Enzyme Activity.
Specifications
Specifications
Supplied as a 0.2μm filtered solution in MES and NaCl.
47 kDa
95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain at 5 μg per lane.
Mouse myeloma cell line,NS0-derived human PCPE-1 protein Met1-Asp449,with a C-terminal 10-His tag
Unconjugated
5118
PCPE-1
20μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70° C as supplied. 3 months, -70° C under sterile conditions after opening.
Yes