Learn More
R&D Systems Human Pentraxin 2/SAP Recombinant Protein
Manufacturer:  R&D Systems 1948SA050/CF
 View more versions of this product
Catalog No. 1948SA050CF
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Lyophilized from a 0.2μm filtered solution in PBS.
25 kDa (monomer)
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Unlabeled
325
Pentraxin 2/SAP
50μg
Noncovalently-linked homopentamer
Yes