Learn More
R&D Systems™ Human Pentraxin 2/SAP Recombinant Protein
Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Binding Activity
Manufacturer:  R&D Systems™ 1948SA050
 View more versions of this product
Catalog No. 1948SA050
Add to cart
This item is not returnable. View return policy
Description
Description
The Recombinant Human Pentraxin 2/SAP Protein is derived from NS0. The Recombinant Human Pentraxin 2/SAP Protein has been validated for the following applications: Binding Activity.
Specifications
Specifications
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
25 kDa (monomer)
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unconjugated
325
Pentraxin 2/SAP
50μg
Noncovalently-linked homopentamer
Yes