Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human Pentraxin 3/TSG-14 Recombinant Protein

Manufacturer:  R&D Systems™ 1826TS025/CF

 View more versions of this product

Catalog No. 1826TS025CF


Add to cart
This item is not returnable. View return policy

Description

Description

Source: Mouse myeloma cell line, NS0-derived, Glu18-Ser381, with a C-terminal 6-His tag
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 41 kDa (monomer)
Formulation: Lyophilized from a 0.2µm filtered solution in PBS.
Entrez Gene ID: 5806
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS.
41 kDa (monomer)
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Multimer consisting of as many as ten non-covalently and covalently linked subunits
Yes
5806
Pentraxin 3/TSG-14
25μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unlabeled
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit