Learn More
R&D Systems Human PGRP-S Recombinant Protein
Manufacturer:  R&D Systems 2590PG050
 View more versions of this product
Catalog No. 2590PG050
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
20.2 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Yes
8993
PGRP-S
50μg
Unlabeled