R&D Systems Human PlGF Recombinant Protein

Manufacturer: R&D Systems 264PG010/CF

 View more versions of this product

Catalog No. 264PG010CF

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    Molecular Weight 14.5 kDa (monomer)
    Name PlGF
    Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
    Source Human
    Formulation Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
    Gene ID (Entrez) 5228
    Quantity 10μg
    Recombinant Yes
    Structural Form Disulfide-linked homodimer
    Conjugate Unlabeled

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit