Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems Human PlGF Recombinant Protein

Manufacturer:  R&D Systems 264PG050/CF

 View more versions of this product

Catalog No. 264PG050CF

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
14.5 kDa (monomer)
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Unlabeled
5228
PlGF
50μg
Disulfide-linked homodimer
Yes
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit