Learn More
R&D Systems Human PlGF Recombinant Protein
Manufacturer:  R&D Systems 264PG010
 View more versions of this product
Catalog No. 264PG010
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
14.5 kDa (monomer)
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Unlabeled
5228
PlGF
10μg
Disulfide-linked homodimer
Yes