Learn More
R&D Systems™ Human Pluripotent Stem Cell Assessment Primer Pair Panel
Catalog No. sc012
Click to view available options
:
1 kit
This item is not returnable. View return policy
This item is not returnable. View return policy
Includes
Primer Pairs, Lyophilized, 375pmol; Positive Control 57, lyophilized dsDNA, 1500ng
For verification of human stem cell pluripotency and differentiation
Verifies differentiation state using PCR
Uses multiple pluripotent and lineage-specific markers
Twelve of the fourteen pluripotent stem cell primer pairs are specific for human cDNA
Efficient and cost-effective
Specifications
Content And Storage Store the unopened product at -20° to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Quantity 1 Kit
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.