Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human Protein Fraction I Protein Fraction

Manufacturer:  R&D Systems™ F37505M

 View more versions of this product

Catalog No. F37505M

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Source: Human cervical epithelial carcinoma cell line, HeLa-derived
Formulation: Supplied as a solution in HEPES.
Regulatory Status: RUO

Specifications

Specifications

Supplied as a solution in HEPES.
5mg
Unlabeled
Protein Fraction I
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit