Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human SIRP gamma/CD172g Fc Chimera Recombinant Protein

Manufacturer:  R&D Systems™ 4486SB050

Catalog No. 4486SB050

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.

This item is not returnable. View return policy

Description

Description

Source: Chinese Hamster Ovary cell line, CHO-derived, Val64-Ser364
Purity: 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 60 kDa
Formulation: Lyophilized from a 0.2µm filtered solution in PBS.
Entrez Gene ID: 55423
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS.
60 kDa
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Chinese Hamster Ovary cell line, CHO-derived Human SIRP gamma (Val64-Ser364) (Val263Ala) (Ser286Leu) Accession # Q9P1W8 IEGRMD Human IgG1 (Pro100-Lys330)
Unlabeled
55423
SIRP gamma/CD172g Fc Chimera
50μg
Disulfide-linked homodimer
Yes
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit