Learn More
R&D Systems Human TGF-beta 1.2 Recombinant Protein
Manufacturer:  R&D Systems 304B3001/CF
 View more versions of this product
Catalog No. 304B3001CF
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
TGF-beta 1.2
1μg
Disulfide-linked heterodimer
Yes
12.8 kDa (TGF-beta1) & 12.7 kDa (TGF-beta2)
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Unlabeled