Learn More
R&D Systems™ Human TGF-beta 1.2 Recombinant Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Supplier:  R&D Systems™ 304B3001
 View more versions of this product
Catalog No. 304B3001
Add to cart
This item is not returnable. View return policy
Description
Description
The Recombinant Human TGF-beta 1.2 Protein is derived from Sf 21 (baculovirus). The Recombinant Human TGF-beta 1.2 Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
TGF-beta 1.2
1μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unconjugated
12.8 kDa (TGF-beta1) & 12.7 kDa (TGF-beta2)
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Spodoptera frugiperda,Sf 21 (baculovirus)-derived human TGF-beta 1.2 protein, (N-terminus) Human TGF-beta 1 (Ala279-Ser390) Accession &Num; P01137 Human TGF-beta 2 (Ala303-Ser414) Accession &Num; P61812 (C-terminus)
Disulfide-linked heterodimer
Yes