Learn More
R&D Systems Human TGF-beta 1 Protein
Manufacturer:  R&D Systems 100B010/CF
 View more versions of this product
Catalog No. 100B010CF
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
TGF-beta 1
10μg
Disulfide-linked homodimer
7040
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Unlabeled