Learn More
R&D Systems™ Human Tryptase epsilon/BSSP-4 Recombinant Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Manufacturer:  R&D Systems™ 5047SE010
Catalog No. 5047SE010
Add to cart
This item is not returnable. View return policy
Description
Description
The Recombinant Human Tryptase epsilon/BSSP-4 Protein is derived from NS0. The Recombinant Human Tryptase epsilon/BSSP-4 Protein has been validated for the following applications: Enzyme Activity.
Specifications
Specifications
Supplied as a 0.2μm filtered solution in Sodium Acetate, NaCl and Glycerol.
32 kDa
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Pro and mature forms
Yes
64063
Tryptase epsilon/BSSP-4
10μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening.
Unlabeled