Learn More
R&D Systems™ Human Ubiquitin Mutant K6R Recombinant Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Supplier: R&D Systems™ UMK6R01M
This item is not returnable.
View return policy
Description
The Recombinant Human Ubiquitin Mutant K6R Protein is derived from E. coli. The Recombinant Human Ubiquitin Mutant K6R Protein has been validated for the following applications: Enzyme Activity.Specifications
Lyophilized from a solution in deionized water. | |
Ubiquitin Mutant K6R | |
E. coli-derived human Ubiquitin protein Contains a Lys-to-Arg substitution at position 6. | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain. |
7314 | |
1mg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |
Provide Content Correction