Learn More
R&D Systems™ Human Ubiquitin Vinyl Sulfone C-Terminal Derivative Recombinant Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity
Manufacturer:  R&D Systems™ U202050
Catalog No. U202050
Add to cart
This item is not returnable. View return policy
Description
Description
The Recombinant Human Ubiquitin Vinyl Sulfone Protein is derived from E. coli. The Recombinant Human Ubiquitin Vinyl Sulfone Protein has been validated for the following applications: Enzyme Activity.
Specifications
Specifications
2.15 mg/mL (250 μM) in 50 mM MES, pH 6.0
8.6 kDa
98%, by HPLC.
E. coli-derived human Ubiquitin protein Contains underivatized and C-terminal VS derivatized protein,quantity is by derivatized content
Vinyl Sulfone
7314
Ubiquitin Vinyl Sulfone C-Terminal Derivative
50μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -70° C as supplied. 3 months, -70° C under sterile conditions after opening.
Yes