Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Human VEGF/PlGF Heterodimer Recombinant Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Supplier: R&D Systems™ 297VP005/CF
This item is not returnable.
View return policy
Description
The Recombinant Human VEGF/PlGF Heterodimer Protein is derived from E. coli. The Recombinant Human VEGF/PlGF Heterodimer Protein has been validated for the following applications: Bioactivity.Specifications
AAM03108 (VEGF) & AAD30179 (PlGF) | |
theoretical_molecular_weight: 19.2 kDa (VEGF) and 14.5 kDa (PIGF) | |
5μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. | |
VEGF/PlGF Heterodimer | |
E. coli-derived human VEGF/PlGF Heterodimer protein, (N-terminus) Human VEGF (Ala27-Arg191) Accession &Num; AAM03108 Human PlGF (Ala21-Arg149) Accession &Num; AAD30179 (C-terminus) | |
Disulfide-linked heterodimer | |
Recombinant |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction