Learn More
R&D Systems™ Human VEGF/PlGF Heterodimer Recombinant Protein
Manufacturer:  R&D Systems™ 297VP005
 View more versions of this product
Catalog No. 297VP005
Add to cart
This item is not returnable. View return policy
Description
Description
Measured in a cell proliferation assay using HUVEC human umbilical vein endothelial cells. Conn, G. et al. (1990) Proc. Natl. Acad. Sci. USA 87:1323. The ED 50 for this effect is 15-75 ng/mL.
Specifications
Specifications
AAM03108 (VEGF) & AAD30179 (PlGF)
28 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
E. coli-derived Human VEGF (Ala27-Arg191) Accession # AAM03108 Human PlGF (Ala21-Arg149) Accession # AAD30179
Disulfide-linked heterodimer
Yes
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
VEGF/PlGF Heterodimer
5μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unlabeled