Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Human XCL1/Lymphotactin Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
$459.36
Specifications
Gene ID (Entrez) | 6375 |
---|---|
Molecular Weight (g/mol) | 10.4 kDa |
Name | XCL1/Lymphotactin |
Quantity | 25μg |
Source | E. coli-derived human XCL1/Lymphotactin protein Val22-Gly114,with an N-terminal Met |
Catalog Number | Mfr. No. | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Formulation | Price | Quantity & Availability | |||||
695LT025CF
|
R&D Systems™
695LT025/CF |
Lyophilized from a 0.2 μm filtered solution in PBS. |
Each for $459.36
|
|
|||||
695LT025
|
R&D Systems™
695LT025 |
Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein. |
Each for $459.36
|
|
|||||
Description
Measured by its ability to chemoattract BaF3 mouse pro-B cells transfected with human XCR1. The ED50 for this effect is 0.0800-0.500 μg/mL.Specifications
6375 | |
XCL1/Lymphotactin | |
E. coli-derived human XCL1/Lymphotactin protein Val22-Gly114,with an N-terminal Met | |
Unconjugated | |
97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
10.4 kDa | |
25μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title