Learn More
Tocris Bioscience™ IHR 1
Potent Smo antagonist
$138.00 - $526.50
Specifications
Product Type Bioactive Small Molecule
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
555410
View Documents
Tocris Bioscience™
5554/10
10 mg
Each for $138.00
Add to cart
555450
View Documents
Tocris Bioscience™
5554/50
50 mg
Each for $526.50
Add to cart
Description
Description
Potent Smo antagonist (IC50 = 7.6 nM). Selectively inhibits Hedgehog signaling over Wnt and Notch signaling pathways. Blocks Smo accumulation in primary cilium in vitro.
Fluorescent Analog also available.
Specifications
Specifications
Bioactive Small Molecule
Safety and Handling
Safety and Handling
Recommended Storage : Store at 4°C
SDS
Product Certifications
Research Use Only. Not For Human or Veterinary Use