Learn More
Tocris Bioscience™ JTE 607 Dihydrochloride Available on GSA/VA Contract for Federal Government customers only.
Cytokine release inhibitor; anti-inflammatory
$254.00 - $1070.00
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
518550 View Documents Tocris Bioscience™
5185/50
Each for $1,070.00
Add to cart
518510 Available on GSA/VA Contract for Federal Government customers only. View Documents Tocris Bioscience™
5185/10
Each for $254.00
Add to cart
Description
Description
Cytokine release inhibitor. Inhibits production of IL-1β, IL-8, IL-6, IL-10 and TNFα (IC50 values are 5.9, 7.3, 8.8, 9.1 and 11.0 nM, respectively) from LPS-stimulated PBMCs. Reduces proinflammatory cytokine-release and attenuates lung permeability in a rat lung injury model. Induces apoptosis in leukemia cells in vitro and prolongs survival in a mouse leukemia model.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Research Use Only. Not For Human or Veterinary Use