R&D Systems Maritoclax, Tocris™

Mcl-1 inhibitor; disrupts Mcl-1-Bim interaction and induces Mcl-1 proteasomal degradation

Manufacturer: R&D Systems 5368/50

 View more versions of this product

Catalog No. 536850

  N/A
Add to Cart

Disclaimers:

Research Use Only. Not For Human or Veterinary Use

  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Specifications

    Physical Form Solid,Solid
    Chemical Name or Material Maritoclax
    Product Type Bioactive Small Molecules
    CAS 1227962-62-0
    Assay Percent Range >97%
    Molecular Formula C22H12Cl4N2O4
    Synonym 1,1'-(4,4',5,5'-Tetrachloro[1,3'-bi-1H-pyrrole]-2,2'-diyl)bis[1-2-hydroxyphenyl)methanone
    Recommended Storage Store at -20°C.
    Quantity 50mg
    Solubility Soluble to 100mM in DMSO and to 50mM in ethanol
    Formula Weight 510.15

    • Exhibits no effect on Bcl-XL-Bim interaction
    • Selectively induces apoptosis in Mcl-1-dependent K562 leukemia cells
    • Sensitizes K562 and Raji cells to ABT-737-induced apoptosis

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit