Learn More
Tocris Bioscience™ Maritoclax
Mcl-1 inhibitor; proapoptotic
Specifications
Product Type Bioactive Small Molecule
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
536810
View Documents
Tocris Bioscience™
5368/10
10 mg This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
536850
View Documents
Tocris Bioscience™
5368/50
50 mg This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description
Description
Mcl-1 inhibitor; disrupts Mcl-1-Bim interaction and induces Mcl-1 proteasomal degradation. Exhibits no effect on Bcl-XL-Bim interaction. Selectively induces apoptosis in Mcl-1-dependent K562 leukemia cells. Sensitizes K562 and Raji cells to ABT-737-induced apoptosis.
Specifications
Specifications
Bioactive Small Molecule
Safety and Handling
Safety and Handling
Recommended Storage : Store at -20°C
SDS
Product Certifications
Research Use Only. Not For Human or Veterinary Use