Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse 4-1BB Ligand/TNFSF9 Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity, Binding Activity

Supplier:  R&D Systems™ 12464L025/CF

 View more versions of this product

Catalog No. 12464L025CF


Add to Cart
This item is not returnable. View return policy

Description

Description

Measured by its ability to co-stimulate IL-2 secretion by mouse T cells in the presence of anti-CD3. The ED50 for this effect is 0.008-0.05 μg/mL in the presence of a Mouse Anti-polyHistidine Monoclonal Antibody (Catalog # MAB050 ). Measured by its ability to bind Recombinant Mouse 4-1BB/TNFRSF9/CD137 Fc Chimera (Catalog # 937-4B ) in a functional ELISA.
Specifications

Specifications

P41274
Lyophilized from a 0.2 μm filtered solution in PBS.
M.W. (Observed): 41-44 kDa, reducing conditions; M.W. (theoretical): 25.7 kDa
Mouse myeloma cell line, NS0-derived mouse 4-1 BB Ligand/TNFSF9 protein Met 10-His tag GGGSGGGSGGGSIEGR Mouse 4-1 BB Ligand (Arg104-Glu309) (N-terminus) Accession # P41274 (C-terminus)
<1.0 EU / 1 μg of the protein by the LAL method.
4-1BB Ligand/TNFSF9
Mouse
Reconstitute at 100 μg/mL in sterile PBS.
4-1 BB Ligand/TNFSF9
Bioactivity, Binding Activity
8744
25 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Tnfsf9
Unconjugated
Recombinant
>95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit