Learn More
R&D Systems Mouse Angiopoietin-like Protein 4/ANGPTL4 Recombinant Protein
Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Inhibition Activity
Manufacturer: R&D Systems 4880AN050
Description
The Recombinant Mouse Angiopoietin-like 4 Protein is derived from CHO. The Recombinant Mouse Angiopoietin-like 4 Protein has been validated for the following applications: Inhibition Activity.This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Lyophilized from a 0.2μm filtered solution in PBS and NaCl. | |
44.2 kDa | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. | |
Chinese Hamster Ovary cell line,CHO-derived mouse Angiopoietin-like Protein 4/ANGPTL4 protein Gln24-Ser410,with a C-terminal 6-His tag | |
Oligomer | |
Yes |
57875 | |
Angiopoietin-like Protein 4/ANGPTL4 | |
50μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.