Learn More
R&D Systems Mouse BAFF/BLyS/TNFSF13B Recombinant Protein
Manufacturer:  R&D Systems 2106BF010
 View more versions of this product
Catalog No. 2106BF010
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications Description & Specifications
Specifications
Lyophilized from a 0.2μm filtered solution in PBS, DTT, EDTA and Tween™ 20 with BSA as a carrier protein.
23.2 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Yes
24099
BAFF/BLyS/TNFSF13B
10μg
Unlabeled