Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Mouse BMPR-IA/ALK-3 Fc Chimera Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
$613.64
Specifications
Gene ID (Entrez) | 12166 |
---|---|
Molecular Weight (g/mol) | 41.5 kDa (monomer) |
Name | BMPR-IA/ALK-3 Fc Chimera |
Quantity | 100μg |
Source | Mouse myeloma cell line,NS0-derived mouse BMPR-IA/ALK-3 protein, (N-terminus) Mouse BMPR-IA (Gln24-Arg152) Accession &Num; Q60607 IEGRMD Human IgG1 (Pro100-Lys330) 6-His tag (C-terminus) |
Catalog Number | Mfr. No. | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Formulation | Price | Quantity & Availability | |||||
437MR100
|
R&D Systems™
437MR100 |
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. |
Each for $613.64
|
|
|||||
437MR100CF
|
R&D Systems™
437MR100/CF |
Lyophilized from a 0.2μm filtered solution in PBS. |
Each for $613.64
|
|
|||||
Description
The Recombinant Mouse BMPR-IA/ALK-3 Fc Chimera Protein is derived from NS0. The Recombinant Mouse BMPR-IA/ALK-3 Fc Chimera Protein has been validated for the following applications: Bioactivity.Specifications
12166 | |
BMPR-IA/ALK-3 Fc Chimera | |
Mouse myeloma cell line,NS0-derived mouse BMPR-IA/ALK-3 protein, (N-terminus) Mouse BMPR-IA (Gln24-Arg152) Accession &Num; Q60607 IEGRMD Human IgG1 (Pro100-Lys330) 6-His tag (C-terminus) | |
Disulfide-linked homodimer | |
Recombinant |
41.5 kDa (monomer) | |
100μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title