Learn More
R&D Systems Mouse CD30 Ligand/TNFSF8 Recombinant Protein
Manufacturer:  R&D Systems 732CL050
 View more versions of this product
Catalog No. 732CL050
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
22 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Yes
21949
CD30 Ligand/TNFSF8
50μg
Unlabeled