Learn More
R&D Systems™ Mouse Cortical Stem Cells (2 x 10e6 cells/vial)
For cortical stem cell expansion and differentiation into neurons, astrocytes and oligodendrocytes
Supplier:  R&D Systems™ NSC002
Catalog No. NSC002
Add to Cart
This item is not returnable. View return policy
Description
Description
  • Have been fully characterized to reduce experimental variation
  • Have been verified to be free of mycoplasma and microbial contamination
Specifications
Specifications
2 x 10e6 cells/vial
Product Suggestions
Product Suggestions