Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Mouse CXCL10/IP-10/CRG-2 Protein
Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Supplier: R&D Systems™ 466CR010/CF
This item is not returnable.
View return policy
Description
Source: E. coli-derived, Ile22-Pro98, with an N-terminal Met
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 8.8 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 15945
Regulatory Status: RUO
Specifications
Lyophilized from a 0.2μm filtered solution in PBS. | |
8.8 kDa | |
10μg | |
Recombinant |
15945 | |
CXCL10/IP-10/CRG-2 | |
Unconjugated | |
97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction