Learn More
R&D Systems Mouse Dkk-2 Recombinant Protein
Manufacturer:  R&D Systems 2435DK010
 View more versions of this product
Catalog No. 2435DK010
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
27.1 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Yes
56811
Dkk-2
10μg
Unlabeled