Learn More
R&D Systems™ Mouse EphA8 Fc Chimera Recombinant Protein
Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Manufacturer:  R&D Systems™ 454A8200
Catalog No. 454A8200
Add to cart
This item is not returnable. View return policy
Description
Description
The Recombinant Mouse EphA8 Fc Chimera Protein is derived from NS0. The Recombinant Mouse EphA8 Fc Chimera Protein has been validated for the following applications: Binding Activity.
Specifications
Specifications
Lyophilized from a 0.2μm filtered solution in MES, NaCl, PEG, CHAPS and Imidazole.
84.2 kDa (monomer)
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
NS0
Disulfide-linked homodimer
Yes
13842
EphA8 Fc Chimera
200μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unlabeled