Learn More
R&D Systems™ Mouse Ephrin-B2 Fc Chimera Recombinant Protein, Biotinylated
Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Binding Activity
Manufacturer:  R&D Systems™ BT496
Catalog No. BT496
Add to cart
This item is not returnable. View return policy
Description
Description
The Recombinant Mouse Ephrin-B2 Fc Chimera Biotinylated Protein is derived from NS0. The Recombinant Mouse Ephrin-B2 Fc Chimera Biotinylated Protein has been validated for the following applications: Binding Activity.
Specifications
Specifications
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
49.6 kDa (monomer)
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse myeloma cell line,NS0-derived mouse Ephrin-B2 protein, (N-terminus) Mouse Ephrin-B2 (Arg27-Ala227) Accession &Num; AAA82934 DIEGRMD Human IgG1 (Pro100-Lys330) 6-His tag (C-terminus)
Disulfide-linked homodimer
Yes
13642
Ephrin-B2 Fc Chimera
25μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Biotin