Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

R&D Systems™ Mouse Exostosin-like 2/EXTL2 Recombinant Protein

Manufacturer:  R&D Systems™ 2536GT020

Catalog No. 2536GT020


This item is not returnable. View return policy

Description

Description

Source: Mouse myeloma cell line, NS0-derived, Asn43-Met330, with an N-terminal 8-His tag
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain at 5 µg per lane.
Molecular Weight: 34 kDa
Formulation: Supplied as a 0.2µm filtered solution in Citric Acid, NaCl, CHAPS and PEG.
Entrez Gene ID: 58193
Regulatory Status: RUO

Specifications

Specifications

Supplied as a 0.2μm filtered solution in Citric Acid, NaCl, CHAPS and PEG.
34 kDa
95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain at 5 μg per lane.
Mouse
<0.1 EU per 1μg of the protein by the LAL method.
Yes
58193
Exostosin-like 2/EXTL2
20μg
Do not freeze. 3 months from date of receipt, 2 to 8° C as supplied. 1 month, 2 to 8° C under sterile conditions after opening.
Unlabeled
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit