Learn More
R&D Systems Mouse Follistatin 288 (FS-288) Recombinant Protein
Manufacturer:  R&D Systems 769FS025/CF
 View more versions of this product
Catalog No. 769FS025CF
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
31 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Yes
14313
Follistatin 288 (FS-288)
25μg
Unlabeled