R&D Systems Mouse GDF-8/Myostatin Propeptide Recombinant Protein

Manufacturer: R&D Systems 1539PG025/CF

 View more versions of this product

Catalog No. 1539PG025CF

  Check Availability
Add to Cart

    Specifications

    Molecular Weight 28.7 kDa
    Name GDF-8/Myostatin Propeptide
    Purity 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
    Quantity 25μg
    Recombinant Yes
    Source Mouse
    Conjugate Unlabeled
    Formulation Lyophilized from a 0.2μm filtered solution in PBS.
    Gene ID (Entrez) 17700

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit