Learn More
R&D Systems™ Mouse Gremlin Recombinant Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Manufacturer:  R&D Systems™ 956GR050
 View more versions of this product
Catalog No. 956GR050
Add to cart
This item is not returnable. View return policy
Description
Description
The Recombinant Mouse Gremlin Protein is derived from NS0. The Recombinant Mouse Gremlin Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
19.6 kDa
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Unlabeled
23892
Gremlin
50μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Yes